Merck
  • Determination of multiple pesticides in fruits and vegetables using a modified quick, easy, cheap, effective, rugged and safe method with magnetic nanoparticles and gas chromatography tandem mass spectrometry.
Determination of multiple pesticides in fruits and vegetables using a modified quick, easy, cheap, effective, rugged and safe method with magnetic nanoparticles and gas chromatography tandem mass spectrometry.
Journal of chromatography. A (2014-08-26)
Yan-Fei Li, Lu-Qin Qiao, Fang-Wei Li, Yi Ding, Zi-Jun Yang, Ming-Lin Wang
RESUMO
Based on a modified quick, easy, cheap, effective, rugged and safe (QuEChERS) sample preparation method with Fe3O4 magnetic nanoparticles (MNPs) as the adsorbing material and gas chromatography-tandem mass spectrometry (GC-MS/MS) determination in multiple reaction monitoring (MRM) mode, we established a new method for the determination of multiple pesticides in vegetables and fruits. It was determined that bare MNPs have excellent function as adsorbent when purified, and it is better to be separated from the extract. The amount of MNPs influenced the clean-up performance and recoveries. To achieve the optimum performance of modified QuEChERS towards the target analytes, several parameters including the amount of the adsorbents and purification time were investigated. Under the optimum conditions, recoveries were evaluated in four representative matrices (tomato, cucumber, orange and apple) with the spiked concentrations of 10 μg kg(-1), 50 μg kg(-1)and 200 μg kg(-1) in all cases. The results showed that the recovery of 101 pesticides ranged between 71.5 and 111.7%, and the relative standard deviation was less than 10.5%. The optimum clean-up system improved the purification efficiency and simultaneously obtained satisfactory recoveries of multiple pesticides, including planar-ring pesticides. In short, the modified QuEChERS method in addition to MNPs used for removing impurities improved the speed of sample pre-treatment and exhibited an enhanced performance and purifying effect.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Chlordimeform, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland