Merck
  • Simultaneous determination of pesticides, polycyclic aromatic hydrocarbons, polychlorinated biphenyls and phthalate esters in human adipose tissue by gas chromatography-tandem mass spectrometry.
Simultaneous determination of pesticides, polycyclic aromatic hydrocarbons, polychlorinated biphenyls and phthalate esters in human adipose tissue by gas chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-05-02)
Na Wang, Deyang Kong, Zhengjun Shan, Lili Shi, Daoji Cai, Yanzhong Cao, Yongming Liu, Guofang Pang
RESUMO
This paper describes a method for the simultaneous determination of 284 environmental contaminants, including 57 pesticides, 15 polycyclic aromatic hydrocarbons (PAHs), 209 polychlorinated biphenyls (PCBs) and 3 phthalate esters (PAEs), in adipose tissue samples. For the first time, a gas chromatography-tandem mass spectrometry (GC-MS/MS) method following a homogenised extraction using acetonitrile and purification by gel permeation chromatography (GPC) was used. Various performance characteristics, such as the limit of detection (LOD), limit of quantification (LOQ), linear range, recovery and precision, were determined for each analyte. The LOD for most analytes was below 0.01 mg/kg. The recoveries and relative standard deviations (RSDs) were determined by spiking untreated samples with the analytes at the LOQ, 2×LOQ and 4×LOQ levels. The average recovery for most pesticides was between 70% and 120% and the precision values, expressed as RSD, were all below 20.4% (n=6). This method may provide an efficient tool for evaluating the extent of exposure to organic contaminants using human adipose tissue.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Chlordimeform, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland