Merck
Cerebral folate deficiency.
Journal of inherited metabolic disease (2010-07-30)
Keith Hyland, John Shoffner, Simon J Heales
RESUMO
Cerebral folate deficiency (CFD) is defined as any neurological syndrome associated with a low cerebrospinal fluid (CSF) concentration of 5-methyltetrahydrofolate (5MTHF) in the presence of normal peripheral folate status. CFD has a wide clinical presentation, with reported signs and symptoms generally beginning at around 4 months of age with irritability and sleep disturbances. These can be followed by psychomotor retardation, dyskinesia, cerebellar ataxia and spastic diplegia. Other signs may include deceleration of head growth, visual disturbances and sensorineural hearing loss. Identification of CFD is achieved by determining 5MTHF concentration in CSF. Once identified, CFD can in many cases be treated by administering oral folinic acid. Supplementation with folic acid is contraindicated and, if used, may exacerbate the CSF 5MTHF deficiency. Generation of autoantibodies against the folate receptor required to transport 5MTHF into CSF and mutations in the folate receptor 1 (FOLR1) gene have been reported to be causes of CFD. However, other mechanisms are probably also involved, as CFD has been reported in Aicardi-Goutiere's and Rett syndromes and in mitochondriopathies. Several metabolic conditions and a number of widely used drugs can also lead to a decrease in the concentration of CSF 5MTHF, and these should be considered in the differential diagnosis if a low concentration of 5MTHF is found following CSF analysis.