Merck
The multifaceted role of MTDH/AEG-1 in cancer progression.
Clinical cancer research : an official journal of the American Association for Cancer Research (2009-09-03)
Guohong Hu, Yong Wei, Yibin Kang
RESUMO
Cancer is the result of the progressive acquisition of multiple malignant traits through the accumulation of genetic or epigenetic alterations. Recent studies have established a functional role of MTDH (Metadherin)/AEG-1 (Astrocyte Elevated Gene 1) in several crucial aspects of tumor progression, including transformation, evasion of apoptosis, invasion, metastasis, and chemoresistance. Overexpression of MTDH/AEG-1 is frequently observed in melanoma, glioma, neuroblastoma, and carcinomas of breast, prostate, liver, and esophagus and is correlated with poor clinical outcomes. MTDH/AEG-1 functions as a downstream mediator of the transforming activity of oncogenic Ha-Ras and c-Myc. Furthermore, MTDH/AEG-1 overexpression activates the PI3K/Akt, nuclear factor kappaB (NFkappaB), and Wnt/beta-catenin signaling pathways to stimulate proliferation, invasion, cell survival, and chemoresistance. The lung-homing domain of MTDH/AEG-1 also mediates the adhesion of tumor cells to the vasculature of distant organs and promotes metastasis. These findings suggest that therapeutic targeting of MTDH/AEG-1 may simultaneously suppress tumor growth, block metastasis, and enhance the efficacy of chemotherapeutic treatments.