Merck
  • Stereospecific and regioselective hydrolysis of cannabinoid esters by ES46.5K, an esterase from mouse hepatic microsomes, and its differences from carboxylesterases of rabbit and porcine liver.
Stereospecific and regioselective hydrolysis of cannabinoid esters by ES46.5K, an esterase from mouse hepatic microsomes, and its differences from carboxylesterases of rabbit and porcine liver.
Biological & pharmaceutical bulletin (2005-09-06)
Kazuhito Watanabe, Tamihide Matsunaga, Toshiyuki Kimura, Tatsuya Funahashi, Satoshi Yamaori, Yukihiro Shoyama, Ikuo Yamamoto
RESUMO
The properties of ES46.5K, an esterase from mouse hepatic microsomes, were compared with those of carboxylesterases from rabbit and porcine liver. The inhibitory profile with a serine hydrolase inhibitor (bis-p-nitrophenylphosphate) and detergents (sodium dodecylsulfate, Emulgen 911) was different between ES46.5K and the carboxylesterases. Bis-p-nitrophenylphosphate (0.1 mM) markedly inhibited the catalytic activity of the carboxylesterases but not that of ES46.5K. Emulgen 911 (0.05-0.25%) inhibited the catalytic activity of the carboxylesterases, whereas the detergent conversely stimulated that of ES46.5K by 150%. The two carboxylesterases catalyzed the hydrolysis of acetate esters of tetrahydrocannabinol (THC) analogues with different side chain lengths (C1-C5), although ES46.5K showed marginal activity only against the acetate of Delta8-tetrahydrocannabiorcol, a methyl side chain derivative of Delta8-THC. ES46.5K hydrolyzed cannabinoid esters stereospecifically and regioselectively. The esterase hydrolyzed 8alpha-acetoxy-Delta9-tetrahydrocannabinol (8alpha-acetoxy-Delta9-THC, 5.62 nmol/min/mg protein), while the enzyme did not hydrolyze 8beta-acetoxy-Delta9-THC, 7alpha-acetoxy-, and 7beta-acetoxy-Delta8-THC at all. In contrast, the carboxylesterases from rabbit and porcine liver hydrolyzed 8beta-acetoxy-Delta9-THC efficiently but not 8alpha-acetoxy-Delta9-THC. ES46.5K hydrolyzed side chain acetoxy derivatives of Delta8-THC at the 3'- and 4'-positions, and a methyl ester of 5'-nor-Delta8-THC-4'-oic acid. The enzyme, however, could not hydrolyze methyl esters of Delta8- and Delta9-THC-11-oic acid, while both carboxylesterases hydrolyzed side chain acetoxy derivatives of Delta8-THC and three methyl esters of THC-oic acids. These differences in stereospecificity and regioselectivity between ES46.5K and carboxylesterases suggest that the configurations of important amino acids for the catalytic activities of these enzymes are different from each other.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid