Merck
Directed disassembly of an interfacial rubisco protein network.
Langmuir : the ACS journal of surfaces and colloids (2007-04-24)
Sagheer A Onaizi, Andrew S Malcolm, Lizhong He, Anton P J Middelberg
RESUMO
We present the first study of the directed disassembly of a protein network at the air-water interface by the synergistic action of a surfactant and an enzyme. We seek to understand the fundamentals of protein network disassembly by using rubisco adsorbed at the air-water interface as a model. We propose that rubisco adsorption at the air-water interface results in the formation of a fishnet-like network of interconnected protein molecules, capable of transmitting lateral force. The mechanical properties of the rubisco network during assembly and disassembly at the air-water interface were characterized by direct measurement of laterally transmitted force through the protein network using the Cambridge interfacial tensiometer. We have shown that, when used individually, either 2 ppm of the surfactant, sodium dodecyl benzyl sulfonate (SDOBS), or 2 ppm of the enzyme, subtilisin A (SA), were insufficient to completely disassemble the rubisco network within 1 h of treatment. However, a combination of 2 ppm SDOBS and 2 ppm SA led to almost complete disassembly within 1 h. Increasing the concentration of SA in the mixture from 2 to 10 ppm, while keeping the SDOBS concentration constant, significantly decreased the time required to completely disassemble the rubisco network. Furthermore, the initial rate of network disassembly using formulations containing SDOBS was surprisingly insensitive to this increase in SA concentration. This study gives insight into the role of lateral interactions between protein molecules at interfaces in stabilizing interfacial protein networks and shows that surfactant and enzyme working in combination proves more effective at disrupting and mobilizing the interfacial protein network than the action of either agent alone.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
HIV Protease Substrate 1