Merck
  • Structural basis for allosteric control of the SERCA-Phospholamban membrane complex by Ca2+ and phosphorylation.
Structural basis for allosteric control of the SERCA-Phospholamban membrane complex by Ca2+ and phosphorylation.
eLife (2021-05-13)
Daniel K Weber, U Venkateswara Reddy, Songlin Wang, Erik K Larsen, Tata Gopinath, Martin B Gustavsson, Razvan L Cornea, David D Thomas, Alfonso De Simone, Gianluigi Veglia
RESUMO
Phospholamban (PLN) is a mini-membrane protein that directly controls the cardiac Ca2+-transport response to β-adrenergic stimulation, thus modulating cardiac output during the fight-or-flight response. In the sarcoplasmic reticulum membrane, PLN binds to the sarco(endo)plasmic reticulum Ca2+-ATPase (SERCA), keeping this enzyme's function within a narrow physiological window. PLN phosphorylation by cAMP-dependent protein kinase A or increase in Ca2+ concentration reverses the inhibitory effects through an unknown mechanism. Using oriented-sample solid-state NMR spectroscopy and replica-averaged NMR-restrained structural refinement, we reveal that phosphorylation of PLN's cytoplasmic regulatory domain signals the disruption of several inhibitory contacts at the transmembrane binding interface of the SERCA-PLN complex that are propagated to the enzyme's active site, augmenting Ca2+ transport. Our findings address long-standing questions about SERCA regulation, epitomizing a signal transduction mechanism operated by posttranslationally modified bitopic membrane proteins.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
4-(Trifluoromethyl)benzyl bromide, 98%