Merck
Horseradish peroxidase-catalyzed polymerization of cardanol in the presence of redox mediators.
Biomacromolecules (2004-01-13)
Keehoon Won, Yong Hwan Kim, Eun Suk An, Yeon Soo Lee, Bong Keun Song
RESUMO
Horseradish peroxidase-catalyzed polymerization of cardanol in aqueous organic solvent was investigated in the presence of a redox mediator. Cardanol is a phenol derivative from a renewable resource mainly having a C15 unsaturated hydrocarbon chain with mostly 1-3 double bonds at a meta position. Unlike soybean peroxidase (SBP), it has been shown that horseradish peroxidase (HRP) is not able to perform oxidative polymerization of phenol derivatives having a bulky meta substituent such as cardanol. For the first time, redox mediators have been applied to enable horseradish peroxidase to polymerize cardanol. Veratryl alcohol, N-ethyl phenothiazine, and phenothiazine-10-propionic acid were tested as a mediator. It is surprising that the horseradish peroxidase-catalyzed polymerization of cardanol took place in the presence of N-ethyl phenothiazine or phenothiazine-10-propionic acid. However, veratryl alcohol showed no effect. FT-IR and GPC analysis of the product revealed that the structure and properties of polycardanol formed by HRP with a mediator were similar to those by SBP. This is the first work to apply a redox mediator to enzyme-catalyzed oxidative polymerization. Our new finding that oxidative polymerization of a poor substrate, which the enzyme is not active with, can take place in the presence of an appropriate mediator will present more opportunities for the application of enzyme-catalyzed polymerization.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Peroxidase, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)