Merck
  • A dual, non-redundant, role for LIF as a regulator of development and STAT3-mediated cell death in mammary gland.
A dual, non-redundant, role for LIF as a regulator of development and STAT3-mediated cell death in mammary gland.
Development (Cambridge, England) (2003-06-18)
Ekaterini A Kritikou, Andrew Sharkey, Kathrine Abell, Paul J Came, Elizabeth Anderson, Richard W E Clarkson, Christine J Watson
RESUMO
STAT3 is the key mediator of apoptosis in mammary gland. We demonstrate here that LIF is the physiological activator of STAT3, because in involuting mammary glands of Lif(-/-) mice, pSTAT3 is absent and the STAT3 target, C/EBPdelta, is not upregulated. Similar to Stat3 knockouts, Lif(-/-) mammary glands exhibit delayed involution, reduced apoptosis and elevated levels of p53. Significantly, Lif(-/-) glands display precocious development during pregnancy, when pSTAT3 is not normally detected. We show that pERK1/2 is significantly reduced in Lif(-/-) glands at this time, suggesting that at this stage LIF mediates its effects through pERK1/2. Inhibition of LIF-mediated ERK1/2 phosphorylation potentiates the proapoptotic effects of STAT3. LIF therefore signals alternately through ERK1/2, then STAT3, to regulate mammary growth and apoptosis.