Merck
Human anti-rhesus D IgG1 antibody produced in transgenic plants.
Transgenic research (2002-06-11)
Thomas Bouquin, Mads Thomsen, Leif Kofoed Nielsen, Trine Hefsgaard Green, John Mundy, Morten Hanefeld Dziegiel
RESUMO
Transgenic plants represent an alternative to cell culture systems for producing cheap and safe antibodies for diagnostic and therapeutic use. To evaluate the functional properties of a 'plantibody', we generated transgenic Arabidopsis plants expressing full-length human IgG1 against the Rhesus D antigen, which is responsible for alloimmunization of RhD- mothers carrying an RhD+ fetus. Anti-RhD extracted from plants specifically reacted with RhD+ cells in antiglobulin technique, and elicited a respiratory burst in human peripheral blood mononuclear cells. Plant-derived antibody had equivalent properties to CHO cell-produced anti-RhD antibody, indicating its potential usefulness in diagnostic and therapeutic programs.