Merck
Placental angiogenesis markers sFlt-1 and PlGF: response to cigarette smoke.
American journal of obstetrics and gynecology (2007-10-02)
Ramkrishna Mehendale, Judith Hibbard, Asgerally Fazleabas, Richard Leach
RESUMO
Excess soluble vascular endothelial growth factor receptor, fms-like tyrosine kinase-1 (sFlt-1), and reduced placental growth factor (PlGF) mediate the genesis of preeclampsia. Cigarette smoking reduces the risk of preeclampsia. We hypothesized that placental secretion of sFlt-1 and PlGF was affected by exposure to cigarette smoke extract. Term placental villous explants were cultured with cigarette smoke extract. Media were analyzed for sFlt-1 and PlGF. Apoptosis was measured by TUNEL staining. Results are reported as sFlt-1 or PlGF picogram/milliliter/milligram wet weight of explant. Exposure to cigarette smoke extract reduced secretion of sFlt-1 in a dose-dependent manner. There was no difference in apoptosis. In contrast with sFlt-1, PlGF did not decline when incubated with cigarette smoke extract. Exposure of placental villous explants to cigarette smoke extract results in a proangiogenic state with reduced sFlt-1 and relative abundance of PlGF. This is the reverse of changes that are seen in preeclampsia and may explain the reduction of preeclampsia in smokers.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
(−)-Nicotine, PESTANAL®, analytical standard