Merck
  • Retro-inverso peptide inhibitor nanoparticles as potent inhibitors of aggregation of the Alzheimer's Aβ peptide.
Retro-inverso peptide inhibitor nanoparticles as potent inhibitors of aggregation of the Alzheimer's Aβ peptide.
Nanomedicine : nanotechnology, biology, and medicine (2016-11-05)
Maria Gregori, Mark Taylor, Elisa Salvati, Francesca Re, Simona Mancini, Claudia Balducci, Gianluigi Forloni, Vanessa Zambelli, Silvia Sesana, Maria Michael, Christos Michail, Claire Tinker-Mill, Oleg Kolosov, Michael Sherer, Stephen Harris, Nigel J Fullwood, Massimo Masserini, David Allsop
RESUMO
Aggregation of amyloid-β peptide (Aβ) is a key event in the pathogenesis of Alzheimer's disease (AD). We investigated the effects of nanoliposomes decorated with the retro-inverso peptide RI-OR2-TAT (Ac-rGffvlkGrrrrqrrkkrGy-NH2) on the aggregation and toxicity of Aβ. Remarkably low concentrations of these peptide inhibitor nanoparticles (PINPs) were required to inhibit the formation of Aβ oligomers and fibrils in vitro, with 50% inhibition occurring at a molar ratio of ~1:2000 of liposome-bound RI-OR2-TAT to Aβ. PINPs also bound to Aβ with high affinity (Kd=13.2-50 nM), rescued SHSY-5Y cells from the toxic effect of pre-aggregated Aβ, crossed an in vitro blood-brain barrier model (hCMEC/D3 cell monolayer), entered the brains of C57 BL/6 mice, and protected against memory loss in APPSWE transgenic mice in a novel object recognition test. As the most potent aggregation inhibitor that we have tested so far, we propose to develop PINPs as a potential disease-modifying treatment for AD.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
Preço e disponibilidade não estão disponíveis no momento.