Merck
  • Ceftaroline: A New Cephalosporin with Activity against Methicillin-Resistant Staphylococcus aureus (MRSA).
Ceftaroline: A New Cephalosporin with Activity against Methicillin-Resistant Staphylococcus aureus (MRSA).
Clinical medicine reviews in therapeutics (2011-07-26)
Christopher Duplessis, Nancy F Crum-Cianflone
RESUMO
Microbial resistance has reached alarming levels, threatening to outpace the ability to counter with more potent antimicrobial agents. In particular, methicillin-resistant Staphylococcus aureus (MRSA) has become a leading cause of skin and soft-tissue infections and PVL-positive strains have been associated with necrotizing pneumonia. Increasing reports of growing resistance to glycopeptides have been noted, further limiting the efficacy of standard antibiotics, such as vancomycin. Ceftaroline is a novel fifth-generation cephalosporin, which exhibits broad-spectrum activity against Gram-positive bacteria, including MRSA and extensively-resistant strains, such as vancomycin-intermediate S. aureus (VISA), heteroresistant VISA (hVISA), and vancomycin-resistant S. aureus (VRSA). In addition to being an exciting new agent in the anti-MRSA armamentarium, ceftaroline provides efficacy against many respiratory pathogens including Streptococcus pneumoniae, Haemophilus influenzae, and Moraxella catarrhalis. Ceftaroline (600 mg intravenously every 12 hours) has been shown effective in phase III studies in the treatment of complicated skin and soft tissue infections and community-acquired pneumonia. To date, this unique antibiotic exhibits a low propensity for inducing resistance and has a good safety profile, although further post-marketing data and clinical experience are needed. In summary, ceftaroline provides an additional option for the management of complex multidrug resistant infections, including MRSA.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ceftaroline fosamil acetate hydrate, ≥98% (HPLC)