Merck
  • A recurrent deletion in the SLC5A2 gene including the intron 7 branch site responsible for familial renal glucosuria.
A recurrent deletion in the SLC5A2 gene including the intron 7 branch site responsible for familial renal glucosuria.
Scientific reports (2016-09-27)
Xiangzhong Zhao, Li Cui, Yanhua Lang, Ting Liu, Jingru Lu, Cui Wang, Sylvie Tuffery-Giraud, Irene Bottillo, Xinsheng Wang, Leping Shao
RESUMO
Familial renal glycosuria (FRG) is caused by mutations in the SLC5A2 gene, which codes for Na+-glucose co-transporters 2 (SGLT2). The aim of this study was to analyze and identify the mutations in 16 patients from 8 families with FRG. All coding regions, including intron-exon boundaries, were analyzed using PCR followed by direct sequence analysis. Six mutations in SLC5A2 gene were identified, including five missense mutations (c.393G > C, p.K131N; c.1003A > G, p.S335G; c.1343A > G, p.Q448R; c.1420G > C, p.A474P; c.1739G > A, p.G580D) and a 22-bp deletion in intron 7 (c.886(-10_-31)del) removing the putative branch point sequence. By the minigene studies using the pSPL3 plasmids, we confirmed that the deletion c.886(-10_-31)del acts as a splicing mutation. Furthermore, we found that this deletion causes exclusion of exon 8 in the SCL5A2 transcript in patients. The mutation c.886(-10_-31)del was present in 5 (62.5%) of 8 families, and accounts for about 37.5% of the total alleles (6/16). In conclusion, six mutations resulting in FRG were found, and the c.886(-10_-31)del may be the high frequency mutation that can be screened in FRG patients with uniallelic or negative SLC5A2 mutations.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
GenElute Blood Genomic DNA Kit, sufficient for 70 purifications