Merck
  • Large-scale application of high-throughput molecular mechanics with Poisson-Boltzmann surface area for routine physics-based scoring of protein-ligand complexes.
Large-scale application of high-throughput molecular mechanics with Poisson-Boltzmann surface area for routine physics-based scoring of protein-ligand complexes.
Journal of medicinal chemistry (2009-04-24)
Scott P Brown, Steven W Muchmore
RESUMO
We apply a high-throughput formulation of the molecular mechanics with Poisson-Boltzmann surface area (htMM-PBSA) to estimate relative binding potencies on a set of 308 small-molecule ligands in complex with the proteins urokinase, PTP-1B, and Chk-1. We observe statistically significant correlation to experimentally measured potencies and report correlation coefficients for the three proteins in the range 0.72-0.83. The htMM-PBSA calculations illustrate the feasibility of procedural automation of physics-based scoring calculations to produce rank-ordered binding-potency estimates for protein-ligand complexes, with sufficient throughput for realization of practical implementation into scientist workflows in an industrial drug discovery research setting.