Merck
Mitochondrial transcription factor A (TFAM) gene variation and risk of late-onset Alzheimer's disease.
Journal of Alzheimer's disease : JAD (2008-04-24)
Victoria Alvarez, Ana I Corao, Cristina Alonso-Montes, Elena Sánchez-Ferrero, Lorena De Mena, Blanca Morales, Mónica García-Castro, Eliecer Coto
RESUMO
Impaired mitochondrial function and an increased number of mutations in mitochondrial DNA (mtDNA) has been found in brains of patients with late-onset Alzheimer's disease (LOAD). The TFAM-gene encodes the mitochondrial transcription factor A, a protein that controls the transcription, replication, damage sensing, and repair of mtDNA. TFAM is on human chromosome region 10q21.1, where a locus for LOAD has been mapped. Our objective was to determine the role of TFAM-gene variation in the risk of LOAD. The seven TFAM coding exons were analysed through single strand conformation analysis and direct sequencing in a cohort of Spanish LOAD-patients and healthy controls. We found four common polymorphisms, two in the flanquing intronic and two in the coding sequences. Polymorphism rs1937 (+35 G/C) was the only missense change (S12T). Genotyping of this polymorphism in 300 LOAD-patients and 183 healthy controls showed a significantly higher frequency of GG-homozygotes in the patients (92% vs. 86%; p=0.04; OR=1.91, 95%CI=1.02-3.50). This suggests that S12 is a risk factor for LOAD in our population. In conclusion, rare variants (mutations) in the TFAM gene were not found in LOAD-patients, but the S12T polymorphism was a moderate risk factor for LOAD in our population.