Merck
An inventory of lysosomal ABC transporters.
FEBS letters (2020-10-25)
Gergely Szakacs, Rupert Abele
RESUMO
ABC transporters fulfill diverse physiological functions in different cellular localizations ranging from the plasma membrane to intracellular membranous compartments. Several ABC transporters have been spotted in the endolysosomal system, which consists of endosomes, autophagosomes, lysosomes, and lysosome-related organelles. In this review, we present an overview of lysosomal ABC transporters including ABCA2, ABCA3, ABCA5, ABCB6, ABCB9, and ABCD4, discussing their trafficking routes, putative substrates, potential physiological functions, and associated diseases. In addition, we offer a critical evaluation of the literature linking ABC transporters to lysosomal drug sequestration, examining pitfalls associated with in vitro models of drug resistance.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Di(2-pyridyl) ketone, 99%