Merck
  • Chemotherapeutic and prophylactic antimalarial drugs induce cell death through mitochondrial-mediated apoptosis in murine models.
Chemotherapeutic and prophylactic antimalarial drugs induce cell death through mitochondrial-mediated apoptosis in murine models.
Drug and chemical toxicology (2018-11-30)
John Oludele Olanlokun, Folashade Abimbola Balogun, Olufunso Olabode Olorunsogo
RESUMO
Malaria is a parasitic disease that has defied many treatment plans. This study was carried out to investigate the host mitochondrial response to malarial infection and selected antimalarial chemotherapy using murine models. The effects of artesunate (ART) and proguanil (PRG) on mitochondrial Permeability Transition (mPT), mitochondrial ATPase (mATPase), level of malondialdehyde (MDA) and activities of antioxidant enzymes; catalase (CAT), superoxide dismutase (SOD), glutathione peroxidase (GPx), Xanthine oxidase (XO), glutathione S-transferase (GST) and reduced glutathione (GSH) were estimated in Plasmodium berghei-infected mice treated with ART and PRG. Besides, apoptotic markers, such as caspases 3, 9 and DNA fragmentation were estimated. Unparasitised (NORMAL) and parasitized but untreated (PU) animals were used as controls. The mPT pore opening fold of 9 (ART), 3 (PRG), and 4 (PU) were observed relative to calcium (23) for in vivo study. In vitro, graded concentrations (20, 40, 80 and 160 μg/mL) of ART gave mPT induction folds of 1, 21, 23 and 25, respectively, relative to calcium (9) while PRG did not have effect in the absence of calcium. In vivo, ART significantly (p < 0.001) enhanced mATPase activity than PRG. The PRG and ART increased the MDA levels in vivo. Oral administration of ART and PRG altered antioxidant enzymes status, Caspases 3 and 9 were significantly activated in PRG-treated groups; there was significant increase in DNA fragmentation in PU and PRG groups compared with the normal control. The results obtained showed that malaria parasite and antimalarial drugs cause mitochondrial-mediated apoptosis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Sodium monododecyl phosphate, AldrichCPR