Merck
  • Inhibition of polymorphic human carbonyl reductase 1 (CBR1) by the cardioprotectant flavonoid 7-monohydroxyethyl rutoside (monoHER).
Inhibition of polymorphic human carbonyl reductase 1 (CBR1) by the cardioprotectant flavonoid 7-monohydroxyethyl rutoside (monoHER).
Pharmaceutical research (2008-05-02)
Vanessa Gonzalez-Covarrubias, James L Kalabus, Javier G Blanco
RESUMO
Carbonyl reductase 1 (CBR1) reduces the anticancer anthracyclines doxorubicin and daunorubicin into the cardiotoxic metabolites doxorubicinol and daunorubicinol. We evaluated whether the cardioprotectant monoHER inhibits the activity of polymorphic CBR1. We performed enzyme kinetic studies with monoHER, CBR1 (CBR1 V88 and CBR1 I88) and anthracycline substrates. We also characterized CBR1 inhibition by the related flavonoids triHER and quercetin. MonoHER inhibited the activity of CBR1 V88 and CBR1 I88 in a concentration-dependent manner. The IC(50) values of monoHER were lower for CBR1 I88 compared to CBR1 V88 for the substrates daunorubicin and doxorubicin (daunorubicin, IC(50)-CBR1 I88 = 164 microM vs. IC(50)-CBR1 V88 = 219 microM; doxorubicin, IC(50)-CBR1 I88 = 37 microM vs. IC(50)-CBR1 V88 = 59 microM; p < 0.001). Similarly, the flavonoids triHER and quercetin exhibited lower IC(50) values for CBR1 I88 compared to CBR1 V88 (p < 0.001). MonoHER acted as a competitive CBR1 inhibitor when using daunorubicin as a substrate Ki = 45 +/- 18 microM. MonoHER acted as an uncompetitive CBR1 inhibitor for the small quinone substrate menadione Ki = 33 +/- 17 microM. The cardioprotectant monoHER inhibits CBR1 activity. CBR1 V88I genotype status and the type of anthracycline substrate dictate the inhibition of CBR1 activity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications