Merck
  • Diterpenes from seeds of Phalaris canariensis and their PTP1B inhibitory activity and hypoglycemic effects in streptozotocin-induced diabetic mice.
Diterpenes from seeds of Phalaris canariensis and their PTP1B inhibitory activity and hypoglycemic effects in streptozotocin-induced diabetic mice.
Journal of Asian natural products research (2019-07-20)
Rosa Martha Perez Gutierrez, Efren Garcia Baez
RESUMO
This present study was to evaluate the protein tyrosine phosphatase 1B (PTP1B) inhibitory activity of nine diterpenes isolated from seeds of Phalaris canariensis, as well as their effect on streptozotocin-nicotinamide-induced type 2 diabetic mice. Their structures were established by spectroscopic analyses. Diterpenes, 1, 4, and 2 exhibited the strongest inhibitory activity on PTP1B with IC50 values of 6.9, 7.3, and 6.5 µM, respectively, The administration of 1-9 showed significant effect on hyperglycemia, among them 1, 4, and 2 reduced fasting glucose levels (55.65%, 54.27%, and 51.22%, respectively). Results revealed that diterpenes performed potential antidiabetic activity via inhibition of PTP1B.[Formula: see text].
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
RK-682, ≥98% (HPLC)