Merck
The circadian clock protects against ferroptosis-induced sterile inflammation.
Biochemical and biophysical research communications (2020-03-03)
Yang Liu, Yuan Wang, Jiao Liu, Rui Kang, Daolin Tang
RESUMO
The circadian clock, a biochemical oscillator, plays a fundamental role in health and diseases. Ferroptosis, a type of regulated cell death driven by oxidative stress, is a prominent feature in iron-induced tissue injury. However, whether an impaired circadian clock contributes to ferroptosis-induced sterile inflammation remains unknown. Here, we show that the circadian transcription factor ARNTL (also known as BMAL1) protects against experimental acute pancreatitis through blocking the ferroptosis-mediated release of HMGB1, a mediator of sterile inflammation. We utilized a Cre/LoxP system to generate mice with a specific depletion of Arntl in the pancreas (Pdx1-Cre;Arntlflox/flox). These Arntl-deficient mice developed l-arginine-induced acute pancreatitis more rapidly than controls, with increased mortality, tissue injury, neutrophil infiltration, and HMGB1 release. In contrast, the administration of liproxstatin-1 (a ferroptosis inhibitor) or anti-HMGB1 neutralizing antibody attenuated the development of acute pancreatitis in the Arntl-deficient mice. Mechanistically, pancreatic ARNTL is a key regulator of the expression of multiple antioxidant or membrane repair systems (e.g., SLC7A11, GPX4, SOD1, TXN, NFE2L2, and CHMP5) to suppress ferroptotic tissue injury. Collectively, these findings uncover a novel link between the circadian clock and ferroptotic response in inflammation and pancreatic injury.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
L-Arginina, reagent grade, ≥98% (HPLC), powder