Merck
  • Overexpression of Rictor protein and Rictor-H. pylori interaction has impact on tumor progression and prognosis in patients with gastric cancer.
Overexpression of Rictor protein and Rictor-H. pylori interaction has impact on tumor progression and prognosis in patients with gastric cancer.
Folia histochemica et cytobiologica (2020-06-27)
Fang Wang, Xiaoqi Lou, Yanfeng Zou, Dingtao Hu, Jiatao Liu, Jie Ning, Yang Jiao, Zuoyang Zhang, Feng Yang, Lulu Fan, Hanqing Yu, Wei Wei, Hua Wang, Guoping Sun
RESUMO
Growing evidence indicates that Rictor (Rapamycin-insensitive companion of mTOR) is overexpressed across several malignancies and associated with poor survival. However, only limited data indicate that Rictor plays a role in gastric cancer (GC). We sought to explore the prognostic value of Rictor in GC and present interaction analysis between Rictor expression and H. pylori status regarding their effects over the prognosis of GC patient. 250 GC tissues and 124 lymph node metastases were collected for the detection of Rictor by immunohistochemistry. Cox regression model was used to assess the association between Rictor expression and patient prognosis. Functional experiments were examined in transfected cells using Rictor siRNA. Additive and multiplicative interactions of Rictor and H. pylori were evaluated. In this study, the positive rate of Rictor was 51.6% (129/150) in GC tissues. Multivariate analyses showed that Rictor was independent unfavorable predictor for OS (HR = 1.554, 95% CI = 1.076-2.244, P = 0.019) and DFS (HR = 1.556, 95% CI = 1.081-2.240, P = 0.017). Patients with upregulated Rictor in the primary tumor and lymph node metastases had the worst prognosis. We observed significant additive and multiplicative interactions between Rictor expression and H. pylori status for OS and DFS (P < 0.05). Our in vitro experiment showed that knockdown of Rictor could suppress cell proliferation, induce apoptosis and inhibit tumor migration and invasion. Our results demonstrate that Rictor, acting as an oncogene, might be a potential prognostic biomarker and therapeutic target in GC. We suggest that Rictor expression and H. pylori status may be a prognostic marker in gastric cancer.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fyb