Merck
  • Ets-1/Elk-1 is a critical mediator of dipeptidyl-peptidase III transcription in human glioblastoma cells.
Ets-1/Elk-1 is a critical mediator of dipeptidyl-peptidase III transcription in human glioblastoma cells.
The FEBS journal (2010-03-20)
Abhay A Shukla, Misti Jain, Shyam S Chauhan
RESUMO
Dipetidyl-peptidase III is a metallopeptidase involved in a number of physiological processes and its expression has been reported to increase with the histological aggressiveness of human ovarian primary carcinomas. Because no information regarding the regulation of its expression was available, experiments were designed to clone, define and characterize the promoter region of the human dipeptidyl-peptidase III (DPP-III) gene. In this study, we cloned a 1038 bp 5'-flanking DNA fragment of the human DPP-III gene for the first time and demonstrated strong promoter activity in this region. Deletion analysis revealed that as few as 45 nucleotides proximal to the transcription start site retained approximately 40% of the activity of the full-length promoter. This promoter lacked the TATA box but contained multiple GC boxes and a single CAAT box. Similarly, two Ets-1/Elk-1-binding motifs are present in the first 25 nucleotides from the transcription start site. Binding of Ets-1/Elk-1 proteins to these motifs was visualized by electrophoretic mobility shift and chromatin immunoprecipitation assays. Mutations of these binding sites abolished not only binding of the Ets protein, but also the intrinsic promoter activity. Increased DNA-binding activity of Ets-1/Elk-1 by v-Ha-ras also augmented the mRNA level and promoter activity of this gene. Similarly, co-transfection of DPP-III promoter-reporter constructs with Ets-1 expression vector led to a significant increase in promoter activity. From these results, we conclude that Ets-1/Elk-1 plays a critical role in transcription of the human DPP-III gene.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dipeptidyl Peptidase III human, recombinant, expressed in Sf9 cells