Merck
Cloning of the Penicillium minioluteum gene encoding dextranase and its expression in Pichia pastoris.
Yeast (Chichester, England) (1996-09-30)
H Roca, B Garcia, E Rodriguez, D Mateu, L Coroas, J Cremata, R Garcia, T Pons, J Delgado
RESUMO
The DEX gene encoding an extracellular dextranase was isolated from the genomic DNA library of Penicillium minioluteum by hybridization using the dextranase cDNA as a probe. Comparison of the gene and cDNA sequences revealed that the DEX gene does not contain introns. Amino acid sequences comparison of P. minioluteum dextranase with other reported dextranases reveals a significant homology (29% identity) with a dextranase from Arthrobacter sp. CB-8. The DEX gene fragment encoding a mature protein of 574 amino acids was expressed in the methylotrophic yeast Pichia pastoris by using the SUC2 gene signal sequence from Saccharomyces cerevisiae under control of the alcohol oxidase-1 (AOX1) promoter. Over 3.2 g/l of enzymatically active dextranase was secreted into the medium after induction by methanol. The yeast product was indistinguishable from the native enzyme in specific activity and the N-terminus of both proteins were identical.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 10-25 units/mg solid