Merck
  • Impact of the anti-inflammatory agent bindarit on the chemokinome: selective inhibition of the monocyte chemotactic proteins.
Impact of the anti-inflammatory agent bindarit on the chemokinome: selective inhibition of the monocyte chemotactic proteins.
European cytokine network (2008-09-09)
Massimiliano Mirolo, Marco Fabbri, Marina Sironi, Annunciata Vecchi, Angelo Guglielmotti, Giorgina Mangano, Giuseppe Biondi, Massimo Locati, Alberto Mantovani
RESUMO
Bindarit is an indazolic derivative that is devoid of any immunosuppressive effects and has no effect on arachidonic acid metabolism. However, it has been proved to have anti-inflammatory activity in a number of experimental diseases, including pancreatitis, arthritis, and lupus nephritis. This therapeutic effect has been associated with its ability to interfere selectively with monocyte recruitment, although the underlying molecular mechanisms are unknown. Here we comprehensively examine the effect of bindarit on the chemokine system, and report that in activated monocytes and endothelial cells, it selectively inhibits the production of the monocyte chemotactic protein subfamily of CC inflammatory chemokines (MCP-1/CCL2, MCP-3/CCL7, MCP-2/CCL8). The capacity of bindarit to inhibit the production of a defined set of related CC chemokines by monocytes and endothelial cells likely underlies the anti-inflammatory activity of this agent in disease. The exploitation of the chemokine system as drug target in inflammatory disease has relied mainly on the development of receptor antagonists and blocking antibodies. Here we report on the use of inhibition of synthesis as a potentially viable and selective approach to modify the chemokine system.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Bindarit, ≥98% (HPLC)