Merck
The human gene encoding GM-CSF is at 5q21-q32, the chromosome region deleted in the 5q- anomaly.
Science (New York, N.Y.) (1985-12-13)
K Huebner, M Isobe, C M Croce, D W Golde, S E Kaufman, J C Gasson
RESUMO
Human granulocyte-macrophage colony-stimulating factor (GM-CSF) is a 22,000-dalton glycoprotein that stimulates the growth of myeloid progenitor cells and acts directly on mature neutrophils. A full-length complementary DNA clone encoding human GM-CSF was used as a probe to screen a human genomic library and isolate the gene encoding human GM-CSF. The human GM-CSF gene is approximately 2.5 kilobase pairs in length with at least three intervening sequences. The GM-CSF gene was localized by somatic cell hybrid analysis and in situ hybridization to human chromosome region 5q21-5q32, which is involved in interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. An established, human promyelocytic leukemia cell line, HL60, contains a rearranged, partially deleted GM-CSF allele and a candidate 5q- marker chromosome, indicating that the truncated GM-CSF allele may reside at the rejoining point for the interstitial deletion on the HL60 marker chromosome.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture