Merck
COPZ1 depletion in thyroid tumor cells triggers type I IFN response and immunogenic cell death.
Cancer letters (2020-02-18)
Tiziana Di Marco, Francesca Bianchi, Lucia Sfondrini, Katia Todoerti, Italia Bongarzone, Elisa Margherita Maffioli, Gabriella Tedeschi, Mara Mazzoni, Sonia Pagliardini, Sandra Pellegrini, Antonino Neri, Maria Chiara Anania, Angela Greco
RESUMO
The coatomer protein complex zeta 1 (COPZ1) represents a non-oncogene addiction for thyroid cancer (TC); its depletion impairs the viability of thyroid tumor cells, leads to abortive autophagy, ER stress, UPR and apoptosis, and reduces tumor growth of TC xenograft models. In this study we investigated the molecular pathways activated by COPZ1 depletion and the paracrine effects on cellular microenvironment and immune response. By comprehensive and target approaches we demonstrated that COPZ1 depletion in TPC-1 and 8505C thyroid tumor cell lines activates type I IFN pathway and viral mimicry responses. The secretome from COPZ1-depleted cells was enriched for several inflammatory molecules and damage-associated molecular patterns (DAMPs). Moreover, we found that dendritic cells, exposed to these secretomes, expressed high levels of differentiation and maturation markers, and stimulated the proliferation of naïve T cells. Interestingly, T cells stimulated with COPZ1-depleted cells showed increased cytotoxic activity against parental tumor cells. Collectively, our findings support the notion that targeting COPZ1 may represent a promising therapeutic approach for TC, considering its specificity for cancer cells, the lack of effect on normal cells, and the capacity to prompt an anti-tumor immune response.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Piroctone olamine, analytical standard, PESTANAL®