Merck
  • WS-7338, new endothelin receptor antagonists isolated from Streptomyces sp. No. 7338. II. Biological characterization and pharmacological characterization of WS-7338 B.
WS-7338, new endothelin receptor antagonists isolated from Streptomyces sp. No. 7338. II. Biological characterization and pharmacological characterization of WS-7338 B.
The Journal of antibiotics (1992-01-01)
S Miyata, M Hashimoto, K Fujie, M Nishikawa, S Kiyoto, M Okuhara, M Kohsaka
RESUMO
WS-7338, produced by Streptomyces sp. No. 7338, was found to be a competitive and specific antagonist against ET-1 receptors in in vitro studies and WS-7338 B is also active in vivo. Furthermore, WS-7338 B was a specific antagonist for vascular ET-1 receptors (ETA receptors) and significantly prevented the accumulation of intracellular inositol 1,4,5-triphosphate (IP3) in endothelin treated rat aorta tissues.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Endothelin Receptor Antagonist W 7338A