Merck
Chemoenzymatic synthesis of neoglycopeptides: application to an alpha-Gal-terminated neoglycopeptide.
The Journal of organic chemistry (2001-04-28)
D Ramos, P Rollin, W Klaffke
RESUMO
A novel methodology for the enzymatic preparation from suitably derivatized oligosaccharides of N-linked neoglycopeptides using the microbial glutaminyl-peptide gamma-glutamyl transferase, transglutaminase (TGase), is described. N-Allyl glycosides of various oligosaccharides were photochemically coupled with cysteamine to yield amino-terminated thioether spacers, which were accepted by transglutaminase to transamidate the side-chain gamma-carboxamide group in the dipeptide Z-Gln-Gly.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Z-Gln-Gly, γ-glutamyl donor substrate