Merck
CSF-1 Overexpression Predicts Poor Prognosis in Upper Tract Urothelial Carcinomas.
Disease markers (2019-10-01)
Wei-Chi Hsu, Yi-Chen Lee, Peir-In Liang, Lin-Li Chang, A-Mei Huang, Hui-Hui Lin, Wen-Jeng Wu, Ching-Chia Li, Wei-Ming Li, Jhen-Hao Jhan, Hung-Lung Ke
RESUMO
Colony-stimulating factor-1 (CSF-1) is a homodimeric glycoprotein. The main role of CSF-1 is as a hematopoietic growth factor that modulates proliferation, differentiation, and survival of macrophages. Moreover, CSF-1 has also been reported to be aberrantly expressed in several human cancers. However, the precise role of CSF-1 in upper tract urothelial carcinomas (UTUC) has not been studied. In this research, we examined the clinical significance of CSF-1 expression in UTUC. One hundred twelve cancer tissue samples of UTUC from patients were included in this study, and the other cohort of 35 UTUC were paired cancer-adjacent normal samples. CSF-1 expression was evaluated by immunohistochemistry, and the association of CSF-1 expression with different clinicopathological variables was analyzed. CSF-1 expression was higher in UTUC than in the normal urothelium (P = 0.005). The CSF-1 expression was primarily localized in the nucleus and was significantly correlated with tumor size (P = 0.04) and patients who had a high stage (P < 0.001), distant metastasis (P = 0.006), recurrence (P = 0.003), and cancer death (P = 0.005). High CSF-1 expression was correlated with poor disease-free survival (P = 0.008) and cancer-specific survival (P = 0.001). Our results also used univariate and multivariable analyses, which found that high CSF-1 expression was an independent predictor of poor disease-free survival (hazard ratio = 2.56; P = 0.007) and cancer-specific survival (hazard ratio = 5.14; P = 0.022). Our findings indicate that the expression of CSF-1 is a potential prognostic marker for predicting patient survival and recurrence in UTUC.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-CSF1 Antibody, clone 2D10, ascites fluid, clone 2D10, from mouse