- Alkylation Reactions of Azodicarboxylate Esters with 4-Alkyl-1,4-Dihydropyridines under Catalyst-Free Conditions.
Alkylation Reactions of Azodicarboxylate Esters with 4-Alkyl-1,4-Dihydropyridines under Catalyst-Free Conditions.
Organic letters (2019-05-31)
Kazunari Nakajima, Yulin Zhang, Yoshiaki Nishibayashi
PMID31145630
RESUMO
Introduction of alkyl groups on azodicarboxylate esters is an important method to prepare alkyl amine derivatives. Herein, we report reactions of 4-alkyl-1,4-dihydropyridines as alkylation reagents with di- tert-butyl azodicarboxylate to prepare alkyl amine derivatives under heating conditions. The alkylation reactions via C-C bond cleavage of the dihydropyridines are achieved in the absence of catalysts and additives.