Merck
  • The antiproliferative effect of C2-ceramide on lung cancer cells through apoptosis by inhibiting Akt and NFκB.
The antiproliferative effect of C2-ceramide on lung cancer cells through apoptosis by inhibiting Akt and NFκB.
Cancer cell international (2014-01-08)
I-Ling Lin, Han-Lin Chou, Jin-Ching Lee, Feng-Wei Chen, Yao Fong, Wei-Chiao Chang, Hurng Wern Huang, Chang-Yi Wu, Wen-Tsan Chang, Hui-Min David Wang, Chien-Chih Chiu
RESUMO
The anticancer effects of ceramide have been reported in many types of cancers but less in lung cancer. In this study, we used C2-ceramide to further investigate its possible anticancer effects and mechanisms on non-small cell lung cancer (NSCLC) H1299 cells. The result of cell proliferation in terms of trypan blue assay showed high dose of C2-ceramide inhibited cell survival after 24 h treatment. The flow cytometry-based assays indicated the effect of apoptosis, chromatin condensation, and G1 arrest in terms of Annexin V/propidium iodide (PI), DAPI, and PI stainings, respectively. Moreover, the decreased protein level of p-Akt, p-NFκB, survivin and cyclin A2 were detected by Western blot assay. Taken together, these results indicated the antiproliferative effect of C2-ceramide is majorly responsible for cell apoptosis in lung cancer H1299 cells.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥97% (TLC), powder