Merck
  • Effects of Controlled Atmosphere on the Storage Quality and Aroma Compounds of Lemon Fruits Using the Designed Automatic Control Apparatus.
Effects of Controlled Atmosphere on the Storage Quality and Aroma Compounds of Lemon Fruits Using the Designed Automatic Control Apparatus.
BioMed research international (2019-07-19)
Yuan Ma, Shaohua Li, Xiaocui Yin, Yage Xing, Hongbin Lin, Qinglian Xu, Xiufang Bi, Cunkun Chen
RESUMO
'Eureka' lemon fruits were stored under four controlled atmosphere- (CA-) combinations at 8°C for 20 days to investigate the effects on weight loss (WL), total soluble solids (TSS), titratable acidity (TA), vitamin C (VC), total phenolic content (TPC), sodium carbonate-soluble pectin (SSP), malondialdehyde (MDA), and volatile compounds. Results showed that the contents of TSS, TA, VC, and SSP in the stored fruits reduced during the storage period, while the WL and MDA increased. Fruit stored under CA2-combination (6 % O2+8 % CO2) showed the lower contents of WL and MDA and the higher content of TSS, TA, TPC, and VC than that of other treated fruits. The main volatile compounds present in the lemons were terpenoids, aldehydes, alcohols, and esters. In addition, both the terpenoid and aldehyde content are substantially higher in lemons exposed to CA2 conditions. In contrast, the alcohols and esters displayed elevated levels in the regular air (RA) stored fruit. In conclusion, CA with the suitable conditions proves to be better than RA as a storage regimen to keep the quality of lemons. These results indicated that the application of 6% O2+8% CO2 CA conditions could maintain the quality of 'Eureka' lemon fruit during the storage time of 20 days and should be the optimal storage environment for postharvest Eureka lemons.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
3-Phenylphenol, 85%