Merck
  • Alniditan, a new 5-hydroxytryptamine1D agonist and migraine-abortive agent: ligand-binding properties of human 5-hydroxytryptamine1D alpha, human 5-hydroxytryptamine1D beta, and calf 5-hydroxytryptamine1D receptors investigated with [3H]5-hydroxytryptamine and [3H]alniditan.
Alniditan, a new 5-hydroxytryptamine1D agonist and migraine-abortive agent: ligand-binding properties of human 5-hydroxytryptamine1D alpha, human 5-hydroxytryptamine1D beta, and calf 5-hydroxytryptamine1D receptors investigated with [3H]5-hydroxytryptamine and [3H]alniditan.
Molecular pharmacology (1996-12-01)
J E Leysen, W Gommeren, L Heylen, W H Luyten, I Van de Weyer, P Vanhoenacker, G Haegeman, A Schotte, P Van Gompel, R Wouters, A S Lesage
RESUMO
Alniditan is a new migraine-abortive agent. It is a benzopyran derivative and therefore structurally unrelated to sumatriptan and other indole-derivatives and to ergoline derivatives. The action of sumatriptan is thought to be mediated by 5-hydroxytryptamine (5-HT)1D-type receptors. We investigated the receptor-binding profile in vitro of alniditan compared with sumatriptan and dihydroergotamine for 28 neurotransmitter receptor subtypes, several receptors for peptides and lipid-derived factors, ion channel-binding sites, and monoamine transporters. Alniditan revealed nanomolar affinity for calf substantia nigra 5-HT1D and for cloned h5-HT1D alpha, h5-HT1D beta and h5-HT1A receptors (Ki = 0.8, 0.4, 1.1, and 3.8 nM, respectively). Alniditan was more potent than sumatriptan at 5-HT1D-type and 5-HT1A receptors. Alniditan showed moderate-to-low or no affinity for other investigated receptors; sumatriptan showed additional binding to 5-HT1F receptors. Dihydroergotamine had a much broader profile with high affinity for several 5-HT, adrenergic and dopaminergic receptors. In signal transduction assays using cells expressing recombinant h5-HT1D alpha, h5-HT1D beta, or h5-HT1A receptors, alniditan (like 5-HT) was a full agonist for inhibition of stimulated adenylyl cyclase (IC50 = 1.1, 1.3, and 74 nM, respectively, for alniditan). Therefore, in functional assays, the potency of alniditan was much higher at 5-HT1D receptors than at 5-HT1A receptors. We further compared the properties of [3H]alniditan, as a new radioligand for 5-HT1D-type receptors, with those of [3H]5-HT in membrane preparations of calf substantia nigra, C6 glioma cells expressing h5-HT1D alpha, and L929 cells expressing h5-HT1D beta receptors. [3H]Alniditan revealed very rapid association and dissociation binding kinetics and showed slightly higher affinity (Kd = 1-2 nM) than [3H]5-HT. We investigated 25 compounds for inhibition of [3H]alniditan and [3H]5-HT binding in the three membrane preparations; Ki values of the radioligands were largely similar, although some subtle differences appeared. Most compounds did not differentiate between 5-HT1D alpha and 5-HT1D beta receptors, except methysergide, ritanserin, ocaperidone, risperidone, and ketanserin, which showed 10-60-fold higher affinity for the 5-HT1D alpha receptor. The Ki values of the compounds obtained with 5-HT1D receptors in calf substantia nigra indicated that these receptors are of the 5-HT1D beta-type. We demonstrated that alniditan is a potent agonist at h5-HT1D alpha and h5-HT1D beta receptors; its properties probably underlie its cranial vasoconstrictive and antimigraine properties.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Alniditan dihydrochloride, ≥98% (HPLC)