Merck
Recombinant thiopeptides containing noncanonical amino acids.
Proceedings of the National Academy of Sciences of the United States of America (2016-03-16)
Xiaozhou Luo, Claudio Zambaldo, Tao Liu, Yuhan Zhang, Weimin Xuan, Chen Wang, Sean A Reed, Peng-Yu Yang, Rongsheng E Wang, Tsotne Javahishvili, Peter G Schultz, Travis S Young
RESUMO
Thiopeptides are a subclass of ribosomally synthesized and posttranslationally modified peptides (RiPPs) with complex molecular architectures and an array of biological activities, including potent antimicrobial activity. Here we report the generation of thiopeptides containing noncanonical amino acids (ncAAs) by introducing orthogonal amber suppressor aminoacyl-tRNA synthetase/tRNA pairs into a thiocillin producer strain of Bacillus cereus .We demonstrate that thiopeptide variants containing ncAAs with bioorthogonal chemical reactivity can be further postbiosynthetically modified with biophysical probes, including fluorophores and photo-cross-linkers. This work allows the site-specific incorporation of ncAAs into thiopeptides to increase their structural diversity and probe their biological activity; similar approaches can likely be applied to other classes of RiPPs.