Merck
  • Characterizing the mode of action of extracellular Connexin43 channel blocking mimetic peptides in an in vitro ischemia injury model.
Characterizing the mode of action of extracellular Connexin43 channel blocking mimetic peptides in an in vitro ischemia injury model.
Biochimica et biophysica acta. General subjects (2016-11-07)
Yeri Kim, Jarred M Griffin, Paul W R Harris, Sin Hang Crystal Chan, Louise F B Nicholson, Margaret A Brimble, Simon J O'Carroll, Colin R Green
RESUMO
Non-selective Connexin43 hemichannels contribute to secondary lesion spread. The hemichannel blocking peptidomimetic Peptide5, derived from the second extracellular loop of the human Connexin43 protein, prevents lesion spread and reduces vascular permeability in preclinical models of central nervous system injury. The molecular mode of action of Peptide5, however, was unknown and is described here. Human cerebral microvascular endothelial cells and APRE-19 cells were used. Scrape loading was used to assess gap junction function and hypoxic, acidic ion-shifted Ringer solution induced ATP release used to assess hemichannel function. Peptide modifications, including amino acid substitutions and truncations, and competition assays were used to demonstrate Peptide5 functional specificity and site of action respectively. Peptide5 inhibits Connexin43 hemichannel-mediated ATP release by acting on extracellular loop two of Connexin43, adjacent to its matching sequence within the protein. Precise sequence specificity is important for hemichannel block, but less so for uncoupling of gap junction channels (seen only at high concentrations). The SRPTEKT motif is central to Peptide5 function but on its own is not sufficient to inhibit hemichannels. Both the SRPTEKT motif and Peptide5 reduce gap junction communication, but neither uncoupling below 50%. Reduced gap junction coupling at high peptide concentrations appears to be relatively non-specific. However, Peptide5 at low concentrations acts upon extracellular loop two of Connexin43 to block hemichannels in a precise, sequence specific manner. The concentration dependent and sequence specific action of Peptide5 supports its development for the treatment of retinal injury and chronic disease, as well as other central nervous system injury and disease conditions.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Connexin43 mimetic peptide 5 trifluoroacetate salt, ≥95% (HPLC)