Merck
Osteogenesis of 3D printed macro-pore size biphasic calcium phosphate scaffold in rabbit calvaria.
Journal of biomaterials applications (2019-01-23)
Fan Liu, Yi Liu, Xinyu Li, Xiaohong Wang, Danni Li, SungMin Chung, Cen Chen, In-Seop Lee
RESUMO
To investigate the osteogenesis of macro-pore sized bone scaffolds, biphasic calcium phosphate scaffolds with accurately controlled macro-pore size (0.8, 1.2, and 1.6 mm) and identical porosity of 70% were fabricated by the 3D printing technology. Eight New Zealand rabbits were selected in the present study, while four 8-mm-diameter calvarial defects were created in each rabbit to place BCP scaffolds with different macro-pore size. The harvested specimens of four and eight weeks were used to evaluate the bone forming ability by micro CT and histological examination. All 3D-printed BCP scaffolds exhibited excellent mechanical properties and had better bone-forming ability than the control at both four and eight weeks. Among them, scaffold with 0.8 mm pore size was superior for initial bone formation and maturation, resulting in the highest value of total bone formation.