Merck
Novel regulatory systems for acetylcholine release in rat striatum and anti-Alzheimer's disease drugs.
Journal of neurochemistry (2019-04-11)
Ikunobu Muramatsu, Junsuke Uwada, Hatsumi Yoshiki, Kiyonao Sada, Kung-Shing Lee, Takashi Yazawa, Takanobu Taniguchi, Matomo Nishio, Takaharu Ishibashi, Takayoshi Masuoka
RESUMO
Regulation of neurotransmitter release in the central nervous system is complex. Here, we investigated regulatory mechanisms for acetylcholine (ACh) release from cholinergic neurons by performing superfusion experiments with rat striatal segments after labelling the cellular ACh pool with [3 H]choline. Electrical stimulation-evoked pronounced [3 H]ACh release from cholinergic neurons. The estimated quantity of [3 H]ACh release per pulse of electrical stimulation was reduced by an increase in stimulus frequency, showing an inverse correlation between release probability of ACh and neuronal excitation. ACh release was also negatively regulated by pre-synaptic muscarinic ACh receptors (mAChRs). The autoinhibition induced by released ACh was predominantly suppressed by the M2 -selective antagonist AF-DX 116, partially inhibited by M3 -selective darifenacin, and minimally by M4 -selective PD 102807. Other subtype-selective antagonists had no effect at subtype-selective concentrations. ACh esterase (AChE) inhibitors (diisopropylfluorophosphate, donepezil and galantamine) at concentrations that mostly inhibit esterase activity reduced [3 H]ACh release, and the reduction was abolished by treatment with atropine. This implies that pre-synaptic autoreceptors are activated more after blockade of ACh hydrolysis, leading to autoinhibition of ACh release and consequent reduction in synaptic ACh concentrations. [3 H]efflux was also enhanced by ACh uptake inhibitors (100 μM hemicholinium-3 and physostigmine), regardless of ACh hydrolysis. This study shows that synaptic ACh concentrations in striatal cholinergic neurons are regulated in a complex manner by many factors such as release probability, pre-synaptic M2 /M3 /M4 mAChRs, AChE and post-synaptic ACh uptake, and provides important information about cholinergic neurotransmission for future exploration of therapeutic strategies for Alzheimer's and other central nervous system diseases. OPEN SCIENCE BADGES: This article has received a badge for *Open Materials* because it provided all relevant information to reproduce the study in the manuscript. The complete Open Science Disclosure form for this article can be found at the end of the article. More information about the Open Practices badges can be found at https://cos.io/our-services/openscience-badges/.