Merck
  • Targeting host metabolism by inhibition of acetyl-Coenzyme A carboxylase reduces flavivirus infection in mouse models.
Targeting host metabolism by inhibition of acetyl-Coenzyme A carboxylase reduces flavivirus infection in mouse models.
Emerging microbes & infections (2019-04-20)
Nereida Jiménez de Oya, William P Esler, Kim Huard, Ayman F El-Kattan, Georgios Karamanlidis, Ana-Belén Blázquez, Priscila Ramos-Ibeas, Estela Escribano-Romero, Andrés Louloudes-Lázaro, Josefina Casas, Francisco Sobrino, Kyle Hoehn, David E James, Alfonso Gutiérrez-Adán, Juan-Carlos Saiz, Miguel A Martín-Acebes
RESUMO
Flaviviruses are (re)-emerging RNA viruses strictly dependent on lipid metabolism for infection. In the search for host targeting antivirals, we explored the effect of pharmacological modulation of fatty acid metabolism during flavivirus infection. Considering the central role of acetyl-Coenzyme A carboxylase (ACC) on fatty acid metabolism, we analyzed the effect of three small-molecule ACC inhibitors (PF-05175157, PF-05206574, and PF-06256254) on the infection of medically relevant flaviviruses, namely West Nile virus (WNV), dengue virus, and Zika virus. Treatment with these compounds inhibited the multiplication of the three viruses in cultured cells. PF-05175157 induced a reduction of the viral load in serum and kidney in WNV-infected mice, unveiling its therapeutic potential for the treatment of chronic kidney disease associated with persistent WNV infection. This study constitutes a proof of concept of the reliability of ACC inhibitors to become viable antiviral candidates. These results support the repositioning of metabolic inhibitors as broad-spectrum antivirals.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-West Nile Virus/Kunjin Antibody, Envelope, clone 3.67G, clone 3.67G, Chemicon®, from mouse
Sigma-Aldrich
PF-05175157, ≥98% (HPLC)