Merck
Fate of the organophosphorus pesticide profenofos in cotton fiber.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2019-01-12)
Mingbo Ma, Suozhuai Dong, Wanhui Jin, Chongqi Zhang, Wenlong Zhou
RESUMO
Pesticides carried by cotton fiber are potential risk for production workers and consumers. Dissipation behaviour of a commonly used cotton pesticide profenofos in cotton fiber during growing period and scouring treatment was investigated. The results showed that profenofos in the fiber from the pre-opened and post-opened bolls both decreased to undetectable amounts 21 days after pesticide application. However, a minority of profenofos was converted to a strongly irritant compound, 4-bromo-2-chlorophenol and retained a non-negligible amount in cotton fiber even after 28 days. Profenofos and its degradation product could be completely removed during the conventional cotton scouring process. The degradation half-time of profenofos in scouring bath was only 3.0 min, and the degradation product was also 4-bromo-2-chlorophenol. Cotton products made of profenofos-carrying fiber are safe; however, the scouring waste should be detoxicated before discharge due to the accumulation of 4-bromo-2-chlorophenol in the scouring bath. These results could be useful for evaluating the risk of cotton fiber from the profenofos applied fields.