Merck
  • Diversity and Ecology of Marine Algicolous Arthrinium Species as a Source of Bioactive Natural Products.
Diversity and Ecology of Marine Algicolous Arthrinium Species as a Source of Bioactive Natural Products.
Marine drugs (2018-12-19)
Young Mok Heo, Kyeongwon Kim, Seung Mok Ryu, Sun Lul Kwon, Min Young Park, Ji Eun Kang, Joo-Hyun Hong, Young Woon Lim, Changmu Kim, Beom Seok Kim, Dongho Lee, Jae-Jin Kim
RESUMO
In our previous study, all Arthrinium isolates from Sargassum sp. showed high bioactivities, but studies on marine Arthrinium spp. are insufficient. In this study, a phylogenetic analysis of 28 Arthrinium isolates from seaweeds and egg masses of Arctoscopus japonicus was conducted using internal transcribed spacers, nuclear large subunit rDNA, β-tubulin, and translation elongation factor region sequences, and their bioactivities were investigated. They were analyzed as 15 species, and 11 of them were found to be new species. Most of the extracts exhibited radical-scavenging activity, and some showed antifungal activities, tyrosinase inhibition, and quorum sensing inhibition. It was implied that marine algicolous Arthrinium spp. support the regulation of reactive oxygen species in symbiotic algae and protect against pathogens and bacterial biofilm formation. The antioxidant from Arthrinium sp. 10 KUC21332 was separated by bioassay-guided isolation and identified to be gentisyl alcohol, and the antioxidant of Arthrinium saccharicola KUC21221 was identical. These results demonstrate that many unexploited Arthrinium species still exist in marine environments and that they are a great source of bioactive compounds.