Merck
  • Injectable in situ dual-crosslinking hyaluronic acid and sodium alginate based hydrogels for drug release.
Injectable in situ dual-crosslinking hyaluronic acid and sodium alginate based hydrogels for drug release.
Journal of biomaterials science. Polymer edition (2019-05-16)
Yufan Zhang, Xian Li, Nan Zhong, Yuanlin Huang, Kewen He, Xu Ye
RESUMO
A series of injectable in situ dual-crosslinking hydrogels (HA/ALG) based on oxidized sodium alginate (oxi-ALG) and hyaluronic acid modified with thiol and hydrazide (HA-SH/CDH) were prepared via hydrazone bonds and disulfide bonds. The chemical structures, morphologies, rheological properties, gelling time, swelling ratio, degradation rate and drug release behavior of hydrogels were investigated. HA/ALG hydrogels exhibited tunable gelling time, rheological properties, swelling ratio and degradation rate with varying precursor concentrations. The gelling time of HA/ALG hydrogels ranged from 157 s to 955 s, the values of yield stress of HA2/ALG2, HA3/ALG3 and HA4/ALG4 hydrogels were 1724, 4349 and 5306 Pa, and the degradation percentage of HA2/ALG2, HA3/ALG3 and HA4/ALG4 hydrogels were about 64%, 51% and 42% after incubating 35 days, respectively. Bovine serum albumin (BSA) was used as a model drug to investigate the drug controlled release properties, and the in vitro cumulative release percentage of BSA from HA2/ALG2, HA3/ALG3 and HA4/ALG4 drug-loaded hydrogels were about 79%, 72% and 69% after 20 days. The series of injectable in situ dual-crosslinking HA/ALG hydrogels could be an attractive candidate for drug delivery system, tissue engineering and regenerative medicine.