Merck
Photochemical Transformations of Dichloroacetamide Safeners.
Environmental science & technology (2019-05-24)
Andrew E Kral, Nicholas C Pflug, Monica E McFadden, Gregory H LeFevre, John D Sivey, David M Cwiertny
RESUMO
Dichloroacetamide safeners are commonly added to commercial chloroacetamide herbicide formulations and widely used worldwide, but their environmental fate has garnered little scrutiny as a result of their classification as "inert" ingredients. Here, we investigated the photolysis of dichloroacetamide safeners to better understand their persistence and the nature of their transformation products in surface waters. High-resolution mass spectrometry (HRMS) and nuclear magnetic resonance (NMR) spectroscopy were used to characterize photoproducts. Of the four commonly used dichloroacetamide safeners, only benoxacor undergoes direct photolysis under simulated natural sunlight ( t1/2 ∼ 10 min). Via a photoinitiated ring closure, benoxacor initially yields a monochlorinated intermediate that degrades over longer irradiation time scales to produce two fully dechlorinated diastereomers and a tautomer, which further photodegrade over several days to a structurally related aldehyde confirmed via NMR. Dichlormid, AD-67, and furilazole were more slowly degraded by indirect photolysis in the presence of the photosensitizers nitrate, nitrite, and humic acid. Reactive entities involved in these reactions are likely hydroxyl radical and singlet oxygen based on the use of selective quenchers. These safeners also directly photolyzed under higher energy ultraviolet (UV) light, suggesting their potential transformation in engineered systems using UV for disinfection. The finding that dichloroacetamide safeners can undergo photolysis in environmental systems over relevant time scales demonstrates the importance of evaluating the fate of this class of "inert" agrochemicals.