Merck
  • Synthesis and Properties of Side-Chain Functionalized Polytetrahydrofuran Derivatives via the Blue-Light Photocatalytic Thiol-Ene Reaction.
Synthesis and Properties of Side-Chain Functionalized Polytetrahydrofuran Derivatives via the Blue-Light Photocatalytic Thiol-Ene Reaction.
Polymers (2019-04-10)
Xiuzhong Zhu, Ting Bai, Zichao Wang, Jie Liu, Xin Min, Tong Wang, Wanbin Zhang, Xiaodong Fan
RESUMO
A series of side-chain functionalized polytetrahydrofuran (PTHF) derivatives were synthesized via the blue-light photocatalytic thiol-ene "click" reaction. Firstly, unsaturated polytetrahydrofuran (UPTHF) as a new unsaturated polyether was synthesized via condensation polymerization of cis-2-butene-1,4-diol and trans-1,4-dibromo-2-butene using potassium hydroxide (KOH) as a catalyst. Then, double bonds in the backbone of UPTHF were modified into different pendant functionality side groups by blue-light photocatalytic thiol-ene "click" reaction using Ru(bpy)₃Cl₂ as a photoredox catalyst, obtaining different side-chain functionalized PTHF derivatives. The structure and the morphology of the side-chain functionalized PTHF derivatives was characterized via Fourier-transform infrared spectra (FTIR), nuclear magnetic resonance (NMR), size exclusion chromatography/multi-angle laser light scattering (SEC/MALLS), and differential scanning calorimeter (DSC). The results showed that the blue-light photocatalytic thiol-ene reaction exhibited high efficiency, and all the unsaturated bonds were modified. Different branch units bestowed different performance of PTHF derivatives; we systematically investigated the thermal properties, pH-triggered and temperature-triggered, self-assembly behaviors of different PTHF derivatives.