Merck
  • Analysis for microbial denitrification and antibiotic resistance during anaerobic digestion of cattle manure containing antibiotic.
Analysis for microbial denitrification and antibiotic resistance during anaerobic digestion of cattle manure containing antibiotic.
Bioresource technology (2019-07-22)
Kaiyu Zhang, Jie Gu, Xiaojuan Wang, Xin Zhang, Ting Hu, Wenya Zhao
RESUMO
This study investigated the effects of tylosin (0, 10, and 100 mg/kg dry weight) on the denitrification genes and microbial community during the anaerobic digestion of cattle manure. N2 emissions were reduced and N2O emissions were increased by 10 mg/kg tylosin. Adding 100 mg/kg tylosin increased the emission of both N2O and N2. The different responses of denitrifying bacteria and genes to tylosin may have been due to the presence of antibiotic resistance genes (ARGs). Network analysis indicated that denitrification genes and ARGs had the same potential host bacteria. intI1 was more important for the horizontal transfer of denitrification genes and ARGs during anaerobic digestion than intI2. The anaerobic digestion of manure containing tylosin may increase nitrogen losses and the associated ecological risk.