Merck
  • Different predation capacities and mechanisms of Harmonia axyridis (Coleoptera: Coccinellidae) on two morphotypes of pear psylla Cacopsylla chinensis (Hemiptera: Psyllidae).
Different predation capacities and mechanisms of Harmonia axyridis (Coleoptera: Coccinellidae) on two morphotypes of pear psylla Cacopsylla chinensis (Hemiptera: Psyllidae).
PloS one (2019-04-24)
Yang Ge, Liu Zhang, Zifang Qin, Yang Wang, Pingping Liu, Shuqian Tan, Zhen Fu, Olivia M Smith, Wangpeng Shi
RESUMO
Pear psylla, Cacopsylla chinensis (Yang & Li) are present as two seasonal morphotypes in pear orchards where they, suck phloem sap, defoliate pear trees, and cause fruit russet. Despite the importance of natural enemies in psyllid control, the interactions between predators and the two seasonal morphotypes of psyllids remain poorly documented. Here we determined the predation efficiencies of the Asian lady beetle, Harmonia axyridis (Pallas) on the two psyllid morphotypes. Predation of H. axyridis on both morphotypes conformed to a Type II functional response: the proportion of consumed psyllids decreased with increasing prey densities. Predation efficiency of H. axyridis against the winterforms increased with temperature when measured from 8 to 25°C. Predation rate on the summerforms was significantly higher than that of the winterforms. This was linked to smaller body size, higher soluble protein level, thinner cuticle, and lower chitin content of summerform psyllids compared to winterform psyllids. Predation capacities of H. axyridis on both morphotypes indicated its potential as a biocontrol agent in psyllids management. Predation efficiency was higher on summerforms, likely due to the difference in body size, exoskeleton fragility, and nutritional value between the two morphotypes. Due to the Type II functional response of H. axyridis to both morphotypes of pear psylla, application of H. axyridis in pear orchards under suitable temperatures could be taken into consideration for suppression of C. chinensis, although further experiments conducted in field conditions are needed to validate our findings.