Merck
  • MRGPRX4 is a G protein-coupled receptor activated by bile acids that may contribute to cholestatic pruritus.
MRGPRX4 is a G protein-coupled receptor activated by bile acids that may contribute to cholestatic pruritus.
Proceedings of the National Academy of Sciences of the United States of America (2019-05-10)
James Meixiong, Chirag Vasavda, Solomon H Snyder, Xinzhong Dong
RESUMO
Patients suffering from cholestasis, the slowing or stoppage of bile flow, commonly report experiencing an intense, chronic itch. Numerous pruritogens are up-regulated in cholestatic patient sera, including bile acids (BAs). Acute injection of BAs results in itch in both mice and humans, and BA-modulating therapy is effective in controlling patient itch. Here, we present evidence that human sensory neuron-expressed Mas-related G protein-coupled receptor X4 (MRGPRX4), an orphan member of the Mrgpr family of GPCRs, is a BA receptor. Using Ca2+ imaging, we determined that pathophysiologically relevant levels of numerous BAs activated MRGPRX4. No mouse Mrgpr orthologs were activated by BAs. To assess the in vivo relevance of BA activation of MRGPRX4, we generated a humanized mouse with targeted expression of MRGPRX4 in itch-encoding sensory neurons. BAs activated MRGPRX4+ sensory neurons at higher levels compared with WT neurons. Compared with control animals, MRGPRX4+ mice scratched more upon acute injection of BAs and in a model of cholestatic itch. Overall, these data suggest that targeting MRGPRX4 is a promising strategy for alleviating cholestatic itch.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tapsigargina, ≥98% (HPLC), solid film
Roche
EagleTaq Universal Master Mix (ROX), sufficient for 100 reactions, sufficient for 500 reactions, sufficient for 5000 reactions, suitable for qPCR, suitable for RT-qPCR