Merck
  • Nirogacestat suppresses RANKL-Induced osteoclast formation in vitro and attenuates LPS-Induced bone resorption in vivo.
Nirogacestat suppresses RANKL-Induced osteoclast formation in vitro and attenuates LPS-Induced bone resorption in vivo.
Experimental cell research (2019-06-19)
Xuzhuo Chen, Xinwei Chen, Zhihang Zhou, Yi Mao, Yexin Wang, Zhigui Ma, Weifeng Xu, An Qin, Shanyong Zhang
RESUMO
Bone resorption, initiated by osteoclasts (OCs), plays an essential role in bone homeostasis. The abnormalities of bone resorption may induce a series of diseases, including osteoarthritis, osteoporosis and aseptic peri-implant loosening. Nirogacestat (PF-03084014, PF), a novel gamma-secretase inhibitor, has been used in phase II clinical trial for treatment of desmoid tumor. However, whether it has the therapeutic effect on abnormal bone resorption remains to be evaluated. In this study, we investigated the role of PF in the regulation of receptor activator of nuclear factor-kB ligand (RANKL)-induced osteoclastogenesis in vitro, and the lipopolysaccharide (LPS)-induced bone resorption in vivo. It was found that PF could suppress the formation of osteoclasts from bone marrow macrophages (BMMs) without causing cytotoxicity, inhibit bone resorption and downregulate the mRNA level of osteoclast-specific markers, including calcitonin receptor (CTR), tartrate resistant acid phosphatase (TRAP), cathepsin K (CTSK), dendritic cell-specific transmembrane protein (Dc-stamp), Atp6v0d2 (V-ATPase d2) and nuclear factor of activated T-cells cytoplasmic 1 (NFATc1). Furthermore, Notch2 signaling, as well as RANKL-induced AKT signaling was significantly inhibited in BMMs. Consistent with in vitro observation, we found that PF greatly ameliorated LPS-induced bone resorption. Taken together, our study demonstrated that PF has a great potential to be used in management of osteolytic diseases.