Merck
  • Tannic acid as a natural antioxidant compound: Discovery of a potent metabolic enzyme inhibitor for a new therapeutic approach in diabetes and Alzheimer's disease.
Tannic acid as a natural antioxidant compound: Discovery of a potent metabolic enzyme inhibitor for a new therapeutic approach in diabetes and Alzheimer's disease.
Journal of biochemical and molecular toxicology (2019-04-12)
Fikret Türkan, Parham Taslimi, Fatma Zerrin Saltan
RESUMO
Multiple studies have been recorded on the synthesis and design of multi-aim anti-Alzheimer molecules. Using dual butyrylcholinesterase/acetylcholinesterase inhibitor molecules has attracted more interest in the therapy for Alzheimer's disease. In this study, a tannic acid compound showed excellent inhibitory effects against acetylcholine esterase (AChE), α-glycosidase, α-amylase, and butyrylcholinesterase (BChE). IC50 values of tannic acid obtained 11.9 nM against α-glycosidase and 3.3 nM against α-amylase, respectively. In contrast, Ki values were found of 50.96 ± 2.18 µM against AChE and 53.17 ± 4.47 µM against BChE. α-Glycosidase inhibitor compounds can be utilized as a novel group of antidiabetic drugs. By competitively decreasing glycosidase activity, these inhibitor molecules help to hamper the fast breakdown of sugar molecules and thereby control the blood sugar level.