Merck
  • Platelet-membrane-camouflaged bismuth sulfide nanorods for synergistic radio-photothermal therapy against cancer.
Platelet-membrane-camouflaged bismuth sulfide nanorods for synergistic radio-photothermal therapy against cancer.
Biomaterials science (2019-07-04)
Yin Chen, Gaomei Zhao, Song Wang, Yongwu He, Songling Han, Changhong Du, Shichao Li, Zhengli Fan, Cheng Wang, Junping Wang
RESUMO
Bismuth-containing nanoparticles (BNPs) are potential enhancers for tumor radiotherapy. Improving the bioavailability and developing synergistic therapeutic regimens benefit the drug transformation of BNPs. In the present study, we prepare a mesoporous silica-coated bismuth nanorod (BMSNR) camouflaged by a platelet membrane (PM). This biomimetic material is termed BMSNR@PM. The PM camouflage enhances the immune escape of the BMSNRs by lowering endocytosis by macrophages in the reticuloendothelial system. Additionally, the PM camouflage strengthens the material tumor-targeting capacity and leads to better radiotherapeutic efficacy compared with bare BMSNRs. Owing to the photothermal effect, BMSNR@PMs alters the cell cycle of 4T1 cancer cells post-treatment with 808 nm near-infrared irradiation (NIR). The proportions of S phase and G2/M phase cells decrease and increase, respectively, which explains the synergistic effect of NIR on BMSNR@PM-based radiotherapy. BMSNR@PMs efficiently eradicates cancer cells by the combined action of photothermal therapy (PTT) and radiotherapy in vivo and markedly improves the survival of 4T1-tumor-bearing mice. The synergistic therapeutic effect is superior to the outcomes of PTT and radiotherapy performed alone. Our study demonstrates a versatile bismuth-containing nanoplatform with tumor-targeting, immune escape, and radiosensitizing functionalities using an autologous cell membrane biomimetic concept that may promote the development of radiotherapy enhancers.